missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CoverWell™ Imaging Chamber Gasket with Adhesive

Catalog No. C24727
Encompass
Change view
Click to view available options
Quantity:
40 gaskets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24727 40 gaskets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24727 Supplier Invitrogen™ Supplier No. C24727

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Incorporate thin, UV-transparent plastic cover, making them ideal for fluorescence microscopy and image analysis

The watertight chambers that form under the covers support and stabilize thick and free-floating specimens, allowing resolution of fine internal structures without the compression or movement artifacts normally seen with ordinary coverslips.

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)

Specifications

Type Imaging Chamber Gasket
Color Clear (Lid)
Lid Material Plastic
Thickness (Metric) 1 mm
Content And Storage Store at room temperature.
Depth (Metric) 1 mm
Diameter (Metric) 20 mm
Shipping Condition Room Temperature
Product Line CoverWell
Closure Color Clear
Quantity 40 gaskets
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.