missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coomassie Fluor™ Orange Protein Gel Stain (Ready-To-Use Solution)

Catalog No. C33250
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33250 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33250 Supplier Invitrogen™ Supplier No. C33250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Staining is complete in less than an hour with no risk of overstaining the gel

Molecular Probes proprietary Coomassie Fluor Orange protein gel stain provides fast, simple, sensitive staining of proteins separated by SDS-PAGE. After electrophoresis, the gel is stained, rinsed and photographed. No separate fixation or destaining steps.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature and protect from light.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Coomassie Fluor Orange
Quantity 1 L
Shipping Condition Room Temperature
Target Molecule Protein
Excitation/Emission 300, 470/570 nm
Product Line Coomassie Fluor
Product Type Protein Gel Stain

For Research Use Only. Not for use in diagnostic procedures.