missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CholEsteryl 4,4-Difluoro-5-(4-Methoxyphenyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Undecanoate (CholEsteryl BODIPY™ 542/563 C11)

Catalog No. C12680
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12680 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12680 Supplier Invitrogen™ Supplier No. C12680

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Orange-red fluorescent cholesteryl ester can be used as tracer of cholesterol transport and rand as general nonexchangeable membrane marker

Cholesteryl esters consist of a fatty acid esterified to the 3β-hydroxyl group of cholesterol. These very nonpolar species are the predominant lipid components of atherosclerosis plaque and low- and high-density lipoporotein (LDL and HDL) cores.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Cholesterol Analogs
Quantity 1 mg
Physical Form Solid
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.