missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Brefeldin A (from Penicillium brefeldianum)

Catalog No. B7450
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B7450 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B7450 Supplier Invitrogen™ Supplier No. B7450
  • ONLINE EXCLUSIVE PRICE
    $208.00
    $170.56 / Each

    Save $37.44

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

missing translation for 'validMainframeMsg'

The fungal metabolite brefeldin A (BFA) has multiple targets in cells and has proven valuable for dissecting the cellular processes, including vesicle formation and kinesin distribution, involved in exporting newly synthesized proteins.

The fungal metabolite brefeldin A (BFA) has multiple targets in cells and has proven valuable for dissecting the cellular processes, including vesicle formation and kinesin distribution, involved in exporting newly synthesized proteins. We are pleased to offer the natural brefeldin A isolated from Penicillium brefeldianum. Exposing cells to BFA causes a distortion in intracellular protein traffic from the ER to the Golgi apparatus and the eventual loss of Golgi apparatus morphology; removal of BFA completely reverses these effects. BFA also alters the morphology of endosomes and lysosomes.

Dissolve in high-quality anhydrous DMSO or ethanol to prepare a stock concentration up to 1 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 5 mg
Shipping Condition Room Temperature
Label Type Unlabeled
Molecular Formula C16H24O4
Physical Form Powder
Product Type Sphingolipid
Molecular Weight (g/mol) 280.36

For Research Use Only. Not for use in diagnostic procedures.