missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAPTA, AM, cell permeant chelator

A cell-permeant chelator, which is a highly selective for Ca2+ over Mg2+

$375.00 - $466.00

Specifications

Content And Storage Store in freezer -5°C to -30°C.
For Use With (Application) Cell Viability and Proliferation
Product Type Cell Permeant Chelator
Dye Type Chelator (Non-Fluorescent)
Shipping Condition Room Temperature
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
B6769
Encompass
View Documents Promotion Details
Invitrogen™
B6769
20 x 1 mg
Each for $466.00
Only null left
 
B1205
Encompass
View Documents Promotion Details
Invitrogen™
B1205
25 mg
Each for $375.00
Only null left
 
Description

Description

  • Can be used to control the level of intracellular Ca2+
  • BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer -5°C to -30°C.
Cell Permeant Chelator
Room Temperature
Cell Viability and Proliferation
Chelator (Non-Fluorescent)
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.