missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAPTA, AM, cell permeant chelator

Catalog No. B1205
Encompass
Change view
Click to view available options
Quantity:
25 mg
20 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1205 25 mg
B6769 20 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1205 Supplier Invitrogen™ Supplier No. B1205
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeant chelator, which is a highly selective for Ca2+ over mg2+

BAPTA, AM is a cell-permeant chelator, which is a highly selective for Ca2+ over Mg2+, and it can be used to control the level of intracellular Ca2+. BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C.
For Use With (Application) Cell Viability and Proliferation
Product Type Cell Permeant Chelator
Dye Type Chelator (Non-Fluorescent)
Quantity 25 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.