missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 1,8-ANS (1-Anilinonaphthalene-8-Sulfonic Acid), high purity

Catalog No. A47
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A47 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A47 Supplier Invitrogen™ Supplier No. A47

A sensitive indicator of protein folding, conformational changes and other processes that modify the exposure of the probe to water.

  • Essentially nonfluorescent in water, only becoming appreciably fluorescent when bound to membranes

Cell Analysis, Cell Structure, Membranes (General) and Lipids

Order Info

Shipping Condition: Room temperature

Specifications

Color Blue
Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type 1,8-ANS
Quantity 100 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids

For Research Use Only. Not for use in diagnostic procedures.