missing translation for 'onlineSavingsMsg'
Learn More

MOF/MYST1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Numéro de catalogue. 501558973
Encompass
Modifier l'affichage
Click to view available options
Quantité:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
501558973 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 501558973 Fournisseur Bethyl Laboratories Inc Code fournisseur A300993AT

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3; MIM 601408), yeast YBF2 and SAS2, and TIP60 (HTATIP; MIM 601409). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.

Spécifications

Antigène MOF/MYST1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugué Unconjugated
Formule phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gène KAT8
Numéro d’acquisition du gène Q9H7Z6
Alias du gène Probable histone acetyltransferase MYST1; MYST protein 1; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; hMOF; MYST1
Symboles de gène KAT8
Espèces hôtes Rabbit
Immunogène Between 417 and C-term
Méthode de purification Antigen affinity chromatography
Quantité 10 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 84148
Espèces cibles Human
Contenu et stockage 4° C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats