Learn More
Modulus Data Systems™ DIFFCOUNT™ III Electronic Differential Tally Counters
Microprocessor-controlled differential cell counter designed specifically for hematology laboratory
Supplier:  Modulus Data Systems™ 10 312
 Afficher plus de versions de ce produit
Numéro de catalogue. 0266948C
Il en reste null
Description
Description
Microprocessor-controlled differential cell counter designed specifically for hematology laboratory
  • Displays all channels simultaneously with large digits on bright backlit LCD display
  • Hermetically sealed switches prevent count anomalies resulting from powdered gloves or other contaminants
  • Compact keyboard with “soft-touch” keys facilitates increased counting speed
  • Key assignment overlays available separately
  • Alarm actuates every 100 counts with optional 100-count lockout
  • Calculates percent and absolute WBC values with count correction
  • Operating temperature range: 0° to 49°C (32° to 120°F)
Model 10-308 feature eight counting keys, plus PERCENT, COUNT, and CLEAR keys. Counting and total displays have three digits
Model 10-312 and 10-312P feature 12 counting keys, plus PERCENT, COUNT, WBC, and CLEAR keys. Each counting channel displays two digits; total displays three digits. Model 10-312P includes a RS-232 printer port.
Spécifications
Identifiants chimiques
LCD
23cm
6cm
15cm
5w
9 in.
2.25 in.
6 in.
50/60Hz
Spécifications
Diff Cell Counter (12-channel)
12.0
23cm
6cm
15cm
110V
Calculates percent or absolute WBC values
LCD
9 in.
2.25 in.
6 in.
50/60Hz
5w
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS