Learn More
Epredia™ Modular Cassette Storage Drawers
Description
Features
- Made from high-impact polystyrene
- Designed with Interlocking ridges on top and bottom to permit stacking to any convenient height
Includes
- Identification labels.
Specifications
Specifications
| Product Type | Cassette Storage Drawer |
| Holds | 250 |