missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Moclobemide

Catalog No. AC465800010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465800010 1 g
AC465802500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465800010 Supplier Thermo Scientific Chemicals Supplier No. 465800010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71320-77-9
Molecular Formula C13H17ClN2O2

Specifications

Quantity 1 g
Melting Point 135.0°C to 138.0°C
Formula Weight 268.74
InChI Key YHXISWVBGDMDLQ-UHFFFAOYSA-N
SMILES ClC1=CC=C(C=C1)C(=O)NCCN1CCOCC1
IUPAC Name 4-chloro-N-[2-(morpholin-4-yl)ethyl]benzamide
Molecular Weight (g/mol) 268.74
Chemical Name or Material Moclobemide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye damage.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

Immediately call a POISON CENTER or doctor/physician.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only