missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Moclobemide

Catalog No. 439550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
439550 50 mg
439510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 439550 Supplier Tocris Bioscience™ Supplier No. 4395/50
Only null left

Reversible MAO-A inhibitor

Reversible monoamine oxidase A (MOA-A) inhibitor.

Chemical Identifiers

CAS 71320-77-9
Molecular Formula C13H17ClN2O2

Specifications

Quantity 50 mg
Formula Weight 268.74
Purity >99%
Molecular Weight (g/mol) 268.741
Chemical Name or Material Moclobemide
Recommended Storage Store at Room Temperature