missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Moclobemide
Description
Reversible monoamine oxidase A (MOA-A) inhibitor.
Spécifications
Spécifications
| Quantity | 10 mg |
| Formula Weight | 268.74 |
| Purity | >99% |
| Molecular Weight (g/mol) | 268.741 |
| Chemical Name or Material | Moclobemide |
Sécurité et manipulation
| Recommended Storage | Store at Room Temperature |