missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MMP-2 Recombinant Protein Antigen

Numéro de catalogue. NBP54667PEP
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
NBP54667PEP 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. NBP54667PEP Fournisseur Novus Biologicals™ Code fournisseur NBP254667PEP
Il en reste null

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human MMP2. The MMP-2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Spécifications

Gene ID (Entrez) 4313
Species Human
Purity >80% by SDS-PAGE and Coomassie blue staining
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Antibody Competition
Gene Symbol MMP2
Molecular Weight (g/mol) 32 kDa
Quantity 100 μL
Regulatory Status RUO
Source E. coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54101. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Afficher plus de résultats Afficher moins de résultats