missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MLR 1023

Catalog No. 458210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
458210 10 mg
458250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 458210 Supplier Tocris Bioscience™ Supplier No. 4582/10
Only null left

Selective allosteric activator of Lyn kinase

Selective allosteric activator of Lyn kinase (EC50 = 63 nM); displays no significant activity against a range of 47 other kinases, including other Src family kinases such as Fyn, Lck and Src kinase. Reduces blood glucose levels without affecting in vivo insulin secretion. Orally bioavailable.

Chemical Identifiers

CAS 41964-07-2
Molecular Formula C11H10N2O2

Specifications

Quantity 10 mg
Formula Weight 202.21
Purity >99%
Molecular Weight (g/mol) 202.213
Chemical Name or Material MLR 1023
Recommended Storage Store at +4°C