accessibility menu, dialog, popup

UserName

CH3OHNON

CAS RN 41964-07-2

IUPAC Name: 5-(3-methylphenoxy)-1,2-dihydropyrimidin-2-one
Synonyms: tolimidone
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: HJQILFPVRNHTIG-UHFFFAOYSA-N
SMILES: CC1=CC=CC(OC2=CNC(=O)N=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results