missing translation for 'onlineSavingsMsg'
Learn More

MLIP Antibody, ProSci

Numéro de catalogue. 16594394
Modifier l'affichage
Click to view available options
Quantité:
20 μg Trial Size
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
16594394 20 μg Trial Size
16919896 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 16594394 Fournisseur ProSci Llc Code fournisseur 67190020

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

Spécifications

Antigène MLIP
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Dilution MLIP antibody can be used for detection of MLIP by Western blot at 1 μg/mL. Antibody can also be used for immunohistochemistry starting at 5 μg/mL. For immunofluorescence start at 20 μg/mL. Antibody validated: Western Blot in human samples; Immunohistochemistry in human samples and Immunofluorescence in human samples. All other applications and species not yet tested.
Formule MLIP Antibody is supplied in PBS containing 0.02% sodium azide.
Numéro d’acquisition du gène NP_612636
Alias du gène MLIP Antibody: C6orf142, C6orf142, Muscular LMNA-interacting protein, Muscular-enriched A-type laminin-interacting protein
Espèces hôtes Rabbit
Méthode de purification MLIP Antibody is affinity chromatography purified via peptide column.
Quantité 20 μg Trial Size
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 90523
Spécifications de test At least three isoforms of MLIP are known to exist.
Espèces cibles Human
Contenu et stockage MLIP antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type de produit Antibody
Forme Liquid
Isotype IgG
Catégorie de recherche Homeostasis
Show More Show Less