missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ mL 334
Description
mL 334 is an Nrf2 activator; inhibits Nrf2/Keap1 interaction (Kd = 1 μM). Promotes Nrf2 nuclear translocation and induces antioxidant response element (ARE) activity in vitro. Cell permeable.
Spécifications
Spécifications
| Chemical Name or Material | (1S,2R)-2-[[(1S)-1-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-3,4-dihydro-2(1H)-isoquinolinyl]carbonyl]cyclohexanecarboxylic acid |
| CAS | 1432500-66-7 |
| Quantity | 5 mg |
| Target | Nrf2 Activators |
| Molecular Formula | C26H26N2O5 |
| Purity | 98% |