missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 120B dihydrochloride

Numéro de catalogue. 489910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
489910 10 mg
489950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 489910 Fournisseur Tocris Bioscience™ Code fournisseur 4899/10
Il en reste null

Novel IKK2-selective inhibitor

Novel, ATP-competitive IKK2-selective inhibitor (IC50 = 60 nM at 50 μM ATP). Exhibits no inhibition at IKK1 (EC50 >100 μM), IKKε (EC50 >100 μM) or a panel of 28 other kinases (EC50 >50 μM). Exhibits antitumor activity in lymphoma-bearing SCID mice. Demonstrates synergistic cytotoxic effects with vincristine (Cat. No. 1257).

Identifiants chimiques

CAS 783348-36-7
Molecular Formula C19H15ClN4O2

Spécifications

Quantity 10 mg
Formula Weight 439.72
Molecular Weight (g/mol) 366.81
Chemical Name or Material ML 120B dihydrochloride
Recommended Storage Store at +4°C