Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
GREENER_CHOICE
Numéro de catalogue. sh3066103
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Port Detail:
4 x Top-mounted Ports and Tubing
Powder Port
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (Metric) Port Detail
SH3066103 200 L 4 x Top-mounted Ports and Tubing
SH3066101 50 L 4 x Top-mounted Ports and Tubing
SH3066102 100 L 4 x Top-mounted Ports and Tubing
SH3068701 50 L Powder Port
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. SH3066103 Fournisseur Thermo Scientific™ Code fournisseur SH30661.03
Il en reste null
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
Spécifications
Capacity (Metric) 200 L
Port Detail 4 x Top-mounted Ports and Tubing
Tubing Line 1 0.25 in. ID Tubing and male/female Luer connectors (Port 1)
Tubing Line 2 0.25 in. ID Tubing and male/female Luer connectors (Port 2)
Tubing Line 3 3/8 in. ID tubing and quick-connect couplings (Port 3)
Tubing Line 4 3/8 in. ID tubing and quick-connect couplings (Port 4)
Tubing Style TPE
Sterility Sterile