missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MitoSOX™ Mitochondrial Superoxide Indicators, for live-cell imaging

Catalog No. M36005
Change view
Click to view available options
Quantity:
1 vial x 50 μg
1 vial x 9 μg
10 vials x 50 μg
2 Vials (1 Green, 1 Red)
5 vials x 9 μg
Excitation/Emission:
∼396/610 nm
∼488/510 nm
488/510 nm, 396/610 nm
Indicator:
MitoSox Green
MitoSox Green, MitoSox Red
MitoSox Red
Color:
Green
Green, Red
Red
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Excitation/Emission Indicator Color
M36005 1 vial x 9 μg ∼488/510 nm MitoSox Green Green
M36006 5 vials x 9 μg ∼488/510 nm MitoSox Green Green
M36008 10 vials x 50 μg ∼396/610 nm MitoSox Red Red
M36007 1 vial x 50 μg ∼396/610 nm MitoSox Red Red
M36009 2 Vials (1 Green, 1 Red) 488/510 nm, 396/610 nm MitoSox Green, MitoSox Red Green, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M36005 Supplier Invitrogen™ Supplier No. M36005

MitoSOX superoxide indicators are novel fluorogenic dyes specifically targeted to mitochondria in live cells. Oxidation of the MitoSOX reagent by mitochondrial superoxide produces bright green or red fluorescence.

MitoSOX Green and MitoSOX Red superoxide indicators are novel fluorogenic dyes specifically targeted to mitochondria in live cells. Oxidation of the MitoSOX reagent by mitochondrial superoxide produces bright green or red fluorescence.

Features of these indicators include:
• Readily oxidized by superoxide but not by other ROS- or RNS-generating systems
• Use for live cell imaging
• Rapidly and selectively targeted to the mitochondria
• MitoSOX Green indicator: absorption/emission maxima of ∼488/510 nm (traditional FITC/GFP filter set)
• MitoSOX Red indicator: absorption/emission maxima of ∼396/610 nm* (custom filter set recommended)

*Note that two excitation peaks may be observed for MitoSOX Red indicator, as it is excited at both 510 nm and 396 nm. While 510 nm will excite the superoxide oxidation product, it can also excite non-specific products, so we recommend using a 396-nm excitation for more selective detection of mito superoxide when using MitoSOX Red indicator (1).

MitoSOX Green indicator is offered as one vial or a pack of five vials. MitoSOX Red indicator is offered as one vial or a pack of ten vials. A variety pack of one vial each of MitoSOX Red and MitSOX Green indicators is also available.

Consult user Manual for solubility instructions.

Quick, easy detection of mitochondrial superoxide in live cells
The production of superoxide by mitochondria can be visualized in fluorescence microscopy using MitoSOX superoxide indicators. They permeate live cells where they selectively target mitochondria. They are rapidly oxidized by superoxide but not by other reactive oxygen species (ROS) and reactive nitrogen species (RNS). The oxidized product is highly fluorescent.

MitoSOX indicators may be used to distinguish artifacts of isolated mitochondrial preparations from direct measurements of superoxide generated in the mitochondria of live cells. They may also provide a valuable tool in the research of agents that modulate oxidative stress in various pathologies. In addition, these indicators have been used in metabolic flux assays using high-content instruments (2). MitoSOX indicators have also been used to detect mitochondrial superoxide using flow cytometery (3).

References
1: Robinson, Kristine M., et al. Selective fluorescent imaging of superoxide in vivo using ethidium-based probes. Proceedings of the National Academy of Sciences 103.41 (2006): 15038-15043.
2: Little, Andrew Charles, et al. High-content fluorescence imaging with the metabolic flux assay reveals insights into mitochondrial properties and functions. Communications biology 3.1 (2020): 1-10.
3: Kauffman, Megan E. et al. MitoSOX-Based Flow Cytometry for Detecting Mitochondrial ROS. Reactive oxygen species (Apex, N.C.) vol. 2,5 (2016): 361-370.

Specifications

Color Green
Quantity 1 vial x 9 μg
Detection Method Fluorescence
Excitation/Emission ∼488/510 nm
Indicator MitoSox Green
Product Line MitoSOX
Are MitoSOX Mitochondrial Superoxide Indicators fixable?

MitoSOX indicator-stained cells should be imaged within 2 hr and are not fixable.

What is the shipping temperature for MitoSOX Mitochondrial Superoxide Indicators, for live-cell imaging?

MitoSOX Mitochondrial Superoxide Indicators, for live-cell imaging are shipped at ambient temperature and should be stored as recommended.

Can I use MitoSOX Mitochondrial Superoxide Indicators (Cat. Nos. M36005, M3006, M36007, M36008, M36009) with other ROS reagents that don't accumulate within the mitochondria?

We do not recommend using MitoSOX Mitochondrial Superoxide Indicators (Cat. Nos. M36005, M3006, M36007, M36008, M36009) with other ROS reagents. Other ROS reagents may compete for radicals in or around the mitochondria limiting the response of the MitoSOX reagents.

Can I store stock solutions of MitoSOX Mitochondrial Superoxide Indicators (Cat. Nos. M36005, M3006, M36007, M36008, M36009) beyond one day?

No. We do not recommend storing MitoSOX Mitochondrial Superoxide Indicators (Cat. Nos. M36005, M3006, M36007, M36008, M36009) stock solutions beyond one day as they are more susceptible to spontaneous oxidation (from radicals in the atmosphere) when in solution compared to storing the reagents in solid form.

Does MitoSOX Green Mitochondrial Superoxide Indicator (Cat. Nos. M36005, M36006) bind to nucleic acids after oxidation?

Upon oxidation, MitoSOX Green Mitochondrial Superoxide Indicator (Cat. Nos. M36005, M36006) does not bind to nucleic acids and unlike MitoSOX Red Mitochondrial Superoxide Indicator (Cat. Nos. M36007, M36008), does not require binding to nucleic acids to enhance its fluorescence.

Is MitoSOX Green Mitochondrial Superoxide Indicator (Cat. No. M36005, M36006) less responsive to other radicals (not superoxide) compared to MitoSOX Red Mitochondrial Superoxide Indicator (Cat. No. M36007, M36008)?

According to Figure 1 of the MitoSOX Green and MitoSOX Red Mitochondrial Superoxide Indicators User Guide, MitoSOX Red shows a slightly higher propensity to oxidation by other radicals compared to MitoSOX Green.