missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C, Thermo Scientific Chemicals

Mitomycin C, CAS # 50-07-7, is a methylazirinopyrroloindoledione compound showing antineoplastic and antibiotic activities. It is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria.

Supplier:  Thermo Scientific Chemicals J63193MA

 View more versions of this product

Catalog No. AAJ63193MA


Only null left

Description

Description

Mitomycin C is used as a chemotherapeutic agent due to its antitumor activity. It is used to treat upper gastro-intestinal cancers. It serves as an antineoplastic agent which inhibits DNA synthesis and induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Mitomycin C is a methylazirinopyrroloindoledione compound isolated from the bacterium Streptomyces caespitosus, and other Streptomyces bacterial species that has antineoplastic and antibiotic activities
• This compound acts as an inhibitor of DNA synthesis, nuclear division, as well as an apoptosis inducer. It can create oxygen radicals, and produce inter-strand DNA crosslinks, inhibiting DNA synthesis
• Mitomycin C shows alkylating activity and mainly targets the guanine nucleoside sequence 5′-CpG-3′. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. Consequently, this interaction prevents the separation of complementary DNA strands, inhibiting DNA replication

Application

• Mitomycin C is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria
• Mitomycin C has strong antineoplastic activity, mainly against Ehrlich ascites tumor cell lines
• This compound can inhibit B and T cells and macrophage proliferation. It can also impair antigen presentation, as well as the secretion of interferon-γ, TNF- α, and IL-2
• This compound is used to generate mitotically inactive feeder cells in cell culture systems, such as the mitotically inactive fibroblasts used in embryonic stem cell systems
• It has been used to treat basal-like cancer cell line in vitro experiments
• It is particularly toxic to hypoxic cells inhibiting RNA and protein synthesis at high concentrations

Specifications

Specifications

Mitomycin C
Powder
360°C
10 mg
MFCD00078109
7231816
Soluble in methanol,water and dimethyl sulfoxide.
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N
5746
334.33
Mitomycin C
50-07-7
Blue to Purple
C15H18N4O5
UN2811
14,6215
NWIBSHFKIJFRCO-WUDYKRTCSA-N
334.332
CHEBI:27504

Chemical Identifiers

50-07-7
334.332
NWIBSHFKIJFRCO-WUDYKRTCSA-N
CHEBI:27504
C15H18N4O5
MFCD00078109
5746
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H351
Fatal if swallowed.
Suspected of causing cancer.

P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H301-H351-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-008-6

RTECSNumber : CN0700000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only