missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C, Thermo Scientific Chemicals

Catalog No. p-7042631
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63193MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63193MA Supplier Thermo Scientific Chemicals Supplier No. J63193MA

Mitomycin C, CAS # 50-07-7, is a methylazirinopyrroloindoledione compound showing antineoplastic and antibiotic activities. It is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria.

Mitomycin C is used as a chemotherapeutic agent due to its antitumor activity. It is used to treat upper gastro-intestinal cancers. It serves as an antineoplastic agent which inhibits DNA synthesis and induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Mitomycin C is a methylazirinopyrroloindoledione compound isolated from the bacterium Streptomyces caespitosus, and other Streptomyces bacterial species that has antineoplastic and antibiotic activities
• This compound acts as an inhibitor of DNA synthesis, nuclear division, as well as an apoptosis inducer. It can create oxygen radicals, and produce inter-strand DNA crosslinks, inhibiting DNA synthesis
• Mitomycin C shows alkylating activity and mainly targets the guanine nucleoside sequence 5′-CpG-3′. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. Consequently, this interaction prevents the separation of complementary DNA strands, inhibiting DNA replication

Application

• Mitomycin C is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria
• Mitomycin C has strong antineoplastic activity, mainly against Ehrlich ascites tumor cell lines
• This compound can inhibit B and T cells and macrophage proliferation. It can also impair antigen presentation, as well as the secretion of interferon-γ, TNF- α, and IL-2
• This compound is used to generate mitotically inactive feeder cells in cell culture systems, such as the mitotically inactive fibroblasts used in embryonic stem cell systems
• It has been used to treat basal-like cancer cell line in vitro experiments
• It is particularly toxic to hypoxic cells inhibiting RNA and protein synthesis at high concentrations

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
MDL Number MFCD00078109
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Type Mitomycin C
Physical Form Powder
Melting Point 360°C
Color Blue to Purple
Quantity 10 mg
UN Number UN2811
Beilstein 7231816
Merck Index 14,6215
Solubility Information Soluble in methanol,water and dimethyl sulfoxide.
Formula Weight 334.33
Chemical Name or Material Mitomycin C
Show More Show Less
Hazard Category H301-H351-H373
Hazard Statement GHS H Statement
H300-H351
Fatal if swallowed.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-008-6
RTECSNumber CN0700000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only