accessibility menu, dialog, popup

UserName

CAS RN 50-07-7

IUPAC Name: [(4S,6S,7R,8S)-11-amino-7-methoxy-12-methyl-10,13-dioxo-2,5-diazatetracyclo[7.4.0.0²,⁷.0⁴,⁶]trideca-1(9),11-dien-8-yl]methyl carbamate
Synonyms: mitomycin c MITC MITO mytozytrex mitomycin (substance) MitocinC mitomycine C MMC MTC mitozytrex
Molecular Weight (g/mol): 334.33
Molecular Formula: C15H18N4O5
InChi Key: NWIBSHFKIJFRCO-WUDYKRTCSA-N
SMILES: CO[C@]12[C@H]3N[C@H]3CN1C1=C([C@H]2COC(N)=O)C(=O)C(N)=C(C)C1=O
Physical Form 
  • (2)
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)