missing translation for 'onlineSavingsMsg'
Learn More

PeproTech MIP-1 gamma Polyclonal Antibody, PeproTech®, Invitrogen™

Numéro de catalogue. 500P1171MG
Encompass
Change view
Click to view available options
Quantity:
100 μg
50 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
500P1171MG 1 mg
500P11750UG 50 μg
500P117100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 500P1171MG Fournisseur PeproTech Code fournisseur 500P1171MG
Il en reste null

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQNSQPRTYK Q. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-1gamma (CCL9/10). Anti-Murine MIP-1gamma (CCL9/10)-specific antibody was purified by affinity chromatography employing an immobilized Murine MIP-1gamma (CCL9/10)matrix. Sandwich ELISA: To detect Murine MIP-1gamma (CCL9/10) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine MIP-1gamma (CCL9/10) (500-P117Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine MIP-1gamma (CCL9/10). Western Blot: To detect Murine MIP-1gamma (CCL9/10) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine MIP-1gamma (CCL9/10) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.

Spécifications

Antigen MIP-1 gamma
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ccl9
Gene Accession No. P51670
Gene Alias C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9
Gene Symbols Ccl9
Host Species Rabbit
Immunogen E.coli-derived Recombinant Murine MIP-1gamma (CCL9/10).
Purification Method Antigen affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20308
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Afficher plus de résultats Afficher moins de résultats