Learn More
MilliporeSigma™ Supelco™ Mininert™ Valve, Screw Thread
Numéro de catalogue. p-7191294
Modifier l'affichage
Click to view available options
Thread Size:
13/425 mm
15/425 mm
18/400 mm
24/400 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Thread Size
111010283 13/425 mm
111020822 15/425 mm
111010255 18/400 mm
111010109 24/400 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 111010283 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 33300
Il en reste null
Mininert™ valves can be used at temperatures up to 105°F.
Benefits include: PTFE construction for inertness. PTFE is useful when working when acids and organic solvents. Good sealing ability for containing most volalite liquids and gases at low pressures. Mininert™ valves can be used at temperatures up to 105°F. 0.036 in. (0.9 mm) opening accepts up to 20-gauge needle. Silicone septa, red.
Spécifications
Color White
Packaging Quantity 12/Pk.
Thread Size 13/425 mm
Material PTFE
Mininert is a registered trademark of Valco Instruments Co. Inc.