missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minigrip™ UV Guard™ UV Protection Bags
Prevent light-sensitive products from deterioration
Supplier: Minigrip™ MGUV3P1212
Description
- Amber
- With Color Line™ and premium zipper
- Film contains UV blocker
- Thickness of 3 mil
- Available in different sizes
Compliance
USP XXII and USP XXIII procedure 66.1.
Chemical Identifiers
| Premium Zipper | |
| Polyethylene | |
| UV Protection Bag |
| Amber | |
| 3 mil |
Specifications
| Premium Zipper | |
| Polyethylene | |
| 12 x 12 in. (30.48 x 30.48cm) | |
| 12 in. |
| Amber | |
| 3 mil | |
| 12 in. | |
| UV Protection Bag |