Learn More
Minigrip™ LabGuard™ Chemotherapy Bags with Absorbent Pad
Used for safe transport of chemotherapy drugs
Supplier: Minigrip™ MGCHEMO1215
Description
Doctors, hospitals, and pharmacies used these secondary storage containers to transport specimen containers from the pharmacy to the patient and return of the empty containers to the pharmacy.
- With reclosable zipper
- Thickness of 4 mil
- Available in two standard sizes
Specifications
| Ziplock | |
| Safe transport of chemotherapy drugs from the pharmacy to the patient and return of the empty containers to the pharmacy | |
| Plastic | |
| 4 mil | |
| 38.10 cm |
| Clear | |
| Absorbent pad, reclosable zipper, yellow and black chemotherapy print | |
| 500 Case | |
| 30.48 cm | |
| Chemotherapy Bag |