Learn More
Cytiva Mini Dialysis Kits
Catalog No. 45001736
Change view
Click to view available options
MWCO:
1 kDa
8 kDa
Capacity (Metric):
250 μL
2 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Capacity (Metric)
45001736 8 kDa 2 mL
45001733 1 kDa 250 μL
45001734 1 kDa 2 mL
45001735 8 kDa 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45001736 Supplier Cytiva Supplier No. 80648432
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
50 tubes with 50 dialysis and 50 standard caps, plus six floats
Offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
  • Especially well-suited for desalting protein samples prior to analysis by isoelectric focusing (IEF) and 2-D electrophoresis
  • Uses disposable dialysis tubes with dialysis membrane incorporated into caps
  • Conical tube bottom maximizes sample recovery
  • Process complete in as little as several hours
  • Choice of molecular weight cut-off and tube size
Specifications
Product Type Mini Dialysis Kit
MWCO 8 kDa
Capacity (Metric) 2 mL
Kit Contents 50 x tube with dialysis cap, 50 x standard cap, 6 x float
Quantity Each
For Use With (Application) Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane