missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Milnacipran hydrochloride

Catalog No. AC466570010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466570010 1 g
AC466572500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466570010 Supplier Thermo Scientific Chemicals Supplier No. 466570010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101152-94-7
Molecular Formula C15H23ClN2O

Specifications

Quantity 1 g
Solubility soluble aq. soln.
Formula Weight 282.81
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key XNCDYJFPRPDERF-PBCQUBLHSA-N
SMILES [H+].[Cl-].CCN(CC)C(=O)[C@@]1(C[C@@H]1CN)C1=CC=CC=C1
IUPAC Name hydrogen (1R,2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide chloride
Molecular Weight (g/mol) 282.81
Chemical Name or Material Milnacipran hydrochloride
Show More Show Less

RUO – Research Use Only