missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Milnacipran hydrochloride

Numéro de catalogue. AC466572500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC466572500 250 mg
AC466570010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC466572500 Fournisseur Thermo Scientific Chemicals Code fournisseur 466572500
Il en reste null

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 101152-94-7
Molecular Formula C15H23ClN2O

Spécifications

Quantity 250 mg
Solubility soluble aq. soln.
Formula Weight 282.81
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key XNCDYJFPRPDERF-PBCQUBLHSA-N
SMILES [H+].[Cl-].CCN(CC)C(=O)[C@@]1(C[C@@H]1CN)C1=CC=CC=C1
IUPAC Name hydrogen (1R,2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide chloride
Molecular Weight (g/mol) 282.81
Chemical Name or Material Milnacipran hydrochloride
Afficher plus de résultats Afficher moins de résultats

RUO – Research Use Only