Learn More
MilliporeSigma Milli-Q™ SQ XL 50 L Tank
Catalog No. TANKSQ050
Change view
Click to view available options
Type: Tank
Catalog No.
TANKSQ050 Type: Tank
1 options
Catalog No. TANKSQ050 Supplier MilliporeSigma Supplier No. TANKSQ050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Store and protect up to 50 L pure RO water for frequent daily usage and direct feed of instruments and equipment
  • 50L Tank storage solution for large vol. of pure RO water produced by Milli-Q™ SR 240XL systems.
  • Optional Gravity valve dispenser and high flow valve available to directly feed lab instruments.
Specifications
Type Tank