missing translation for 'onlineSavingsMsg'
Learn More

Mifepristone 98.0+%, TCI America™

Catalog No. M17325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17325G 5 g
M17321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17325G Supplier TCI America Supplier No. M17325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84371-65-3
Molecular Formula C29H35NO2
Molecular Weight (g/mol) 429.604
InChI Key VKHAHZOOUSRJNA-GCNJZUOMSA-N
Synonym mifepristone, mifeprex, mifegyne, mifepriston, corlux, mifepristona, mifepristonum, korlym, mifepristonum latin, mifepristona spanish
PubChem CID 55245
ChEBI CHEBI:50692
IUPAC Name (8S,11R,13S,14S,17S)-11-[4-(dimethylamino)phenyl]-17-hydroxy-13-methyl-17-prop-1-ynyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES CC#CC1(CCC2C1(CC(C3=C4CCC(=O)C=C4CCC23)C5=CC=C(C=C5)N(C)C)C)O

Specifications

Melting Point 195°C
Quantity 5 g
Formula Weight 429.60
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mifepristone
RTECSNumber KG2955000
TSCA No
Recommended Storage Refrigerator