Learn More
Applied Biosystems™ MicroSEQ™ ID Ultra Sequencing Clean-up Plates Kit
Description
The MicroSEQ™ ID Ultra Sequencing Clean-up Plates Kit is used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel-filtration matrix used in the microplate cartridges provides a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions. MicroSEQ ID Ultra Sequencing Clean-up plates can be stored at ambient temperate for up to 4 weeks. The MicroSEQ ID Ultra Sequencing Clean-up Plates Kit offers the same or better sequence quality results as the MicroSEQ™ ID Sequencing Clean-up Plates.
The MicroSEQ ID Ultra Sequencing Clean-up plates can be used downstream of the following sequencing kits:
- MicroSEQ™ 500 16S rDNA Sequencing Kit
- MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
- MicroSEQ™ D2 rDNA Fungal Sequencing Kit
This kit provides two 96-well microplate cartridges ideal for high-throughput microbial-identification workflows. The end products can be used directly for sequencing on any Applied Biosystems™ genetic analyzers.
Specifications
Specifications
| Content And Storage | • 2 x 96-well reaction plates • 2 x 96-well plate lids • 2 x 96-well waste plates • 2 x 96-well semi-skirted capillary plates |
| Format | 96-well Plate |
| Sample Type | Sequencing Products |
| For Use With (Application) | Sequencing |
| High-throughput Compatibility | High-throughput Compatible |
| No. of Samples | 192 Samples |
| Product Line | MicroSEQ™ ID |
| Quantity | 192 samples |
| Target | Fluorescent Dye Removal |
Frequently Asked Questions (FAQs)
The product is guaranteed stable for one year when stored properly at 4 degrees C.
Yes. The kit contents are stable when stored in their original packaging at room temperature for up to 4 weeks. However, the recommended storage condition is at 4 degrees C.
Unfortunately, frozen plates cannot be used. Continued use will result in total sample loss.
The MicroSEQ ID Ultra Sequencing Clean-up Plates Kit is sufficient for 192 sequencing samples.
Here the kits with the catalog numbers:
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)
- MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No.A33245)
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)