missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Assay/Reader Microplate

Catalog No. E951040145
Change view
Click to view available options
Color:
Gray
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
No. of Wells:
384
96
Well Color:
Black
White
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape No. of Wells Well Color
E951040145 Gray Round Bottom (U-well) 96 White
05414244 White Flat Bottom (F-well) 96 -
05414243 Gray Flat Bottom (F-well) 96 -
E951040308 Gray Conical Bottom (V-well) 96 White
E951040102 White Round Bottom (U-well) 96 Black
E951040260 White Conical Bottom (V-well) 96 Black
E951040481 White Conical Bottom (V-well) 384 Black
E951040503 Gray Conical Bottom (V-well) 384 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. E951040145 Supplier Eppendorf™ Supplier No. 951040145

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.

  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
  • Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
  • All plates are optimized for minimal autofluorescence and autoluminescence.
  • Available in lot-certified PCR clean purity grade.
  • Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.

Specifications

Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 16 Plates
Color Gray
Product Type Microplate
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Round Bottom (U-well)
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 80 Pk.
Well Color White
Show More Show Less