missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Plate Sealing Mats, 7 mm Diameter, Round, Level 2 High-Throughput Applications

Use SureSTART™ WebSeal™ 96-Well Plate Sealing Mats, 7 mm Diameter, Round, Performance Level 2 for high-throughput chromatography analysis. They are specification certified and conform to ANSI-SLAS 4-2004 standards.

Spécifications

Sterility Non-sterile
No. of Wells 96
Plug Diameter 7 mm
For Use With (Application) HPLC, UHPLC, GC
Product Line SureSTART WebSeal Level 2
voir les spécifications

Produits 8
Numéro de catalogue Numéro du manufacturier. Pre-slit Material Format No. per Pack Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Pre-slit Material Format No. per Pack Prix Quantity & Availability  
03252163
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M149
No Silicone Step Base, Small Penetration Area 5 Mats N/A
Il en reste null
 
03255000
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M105
No Silicone Flat Base, Large Penetration Area 5 N/A
Il en reste null
 
03252164
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M113
Yes Silicone Step Base, Small Penetration Area 5 Mats N/A
Il en reste null
 
03252162
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M116
Yes Silicone Flat Base, Large Penetration Area 5 Mats N/A
Il en reste null
 
03252170
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M111
No Silicone, PTFE Flat Base 5 Mats N/A
Il en reste null
 
03252171
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M112
Yes Silicone, PTFE Flat Base 5 Mats N/A
Il en reste null
 
03255001
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M179
No EVA Domed Base 100 Mats N/A
Il en reste null
 
01213156
Afficher les documents Détails de la promotion
Thermo Scientific™
60180M187
No Silicone Rubber Flat Base, Large Penetration Area 50 N/A
Il en reste null
 
Description

Description

Thermo Scientific™ SureSTART™ Level 2 WebSeal™ 7 mm Diameter Round 96-Well Plate Sealing Mats are designed to ensure high quality data with an uninterrupted workflow for high-throughput applications using GC and HPLC/UHPLC and are ideal for regulated labs that use water and polar solvents.

These 96-well plate sealing mats are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards.

Performance Level 2 Mats – for High-Throughput Applications 

  • Ideal for non-regulated labs using water and polar solvents
  • GC-MS and LC-MS tested
  • Suitable for HPLC/UHPLC,QA/QC, and sensitive applications
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, fluorescence, and ELSD
  • Eliminates cross contamination of samples
  • Resists coring and tearing
  • Superior resealability after multiple injections
  • WebSeal mats are manufactured from EVA (Ethylene-vinyl acetate) or pure silicone with or without a PTFE coating
  • Silicone mats are dry heat autoclavable with excellent chemical compatibility, and can withstand low temperatures down to -80°C

Reduce the risk of mis-injection – To seal your plates correctly, it’s important to use the right mat

  • Silicone Mats – Using a silicone mat with the correct profile reduces sample and solvent evaporation during your workflows, provides better consistency, and is ideal for long-term storage.
  • Pre-slit Mats – Pre-slit mats are effective at reducing mis-injections and can be used again for confirmatory runs on the same plates without solvent loss. Their slits allow the use of pipette tips as well as multichannel pipettes.
  • Adhesive Seals – Adhesive seals are effective as a one-time seal, but cannot be used for repeat runs. The exception is our three-layer re-sealing tape without glue at the cavity area.

Specification Certified  

  • All Thermo Scientific SureSTART WebSeal mats are dimensionally verified 

Lot Traceable

  • 100% traceable through the manufacturing process 
  • Certificate of Conformance available by lot number upon request 

About Thermo Scientific WebSeal Well Plates & Mats

Thermo Scientific WebSeal plates and mats are available in three performance levels:

  • Level 1 products are for everyday applications and are suitable for non-regulated labs using water and polar solvents
  • Level 2 products are for high-throughput applications and are suitable for regulated labs using water and polar solvents
  • Level 3 products are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents
Spécifications

Spécifications

Non-sterile
7 mm
SureSTART WebSeal Level 2
Yes
96
HPLC, UHPLC, GC
None
Round
Videos
FDS
Documents

Documents

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.