Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Catalog No. 361013375
Change view
Click to view available options
For Use With (Equipment):
HM 525
HM 550 and 560
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
361013375 HM 525
1407088 HM 550 and 560
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361013375 Supplier Epredia™ Supplier No. 524490
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
Specifications
Product Type Heat Extractor
For Use With (Equipment) HM 525