missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
GREENER_CHOICE

For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.

Supplier:  Greiner Bio-One 705074

 Afficher plus de versions de ce produit

Numéro de catalogue. 07000782


Il en reste null

Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Individual wells can be pipetted without leaving residue
  • Flat bottom enables precise optical measurements
  • Suitable for transmission measurements
  • Specifica
Spécifications

Identifiants chimiques

Single break
8 x 12
96
Curved Edges, Flat Bottom (C-well)
Colorimetric immunoassay
127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.)
Strip Plate
Polystyrene
Non-sterile
Yes
Yes

Spécifications

Single break
Strip Plate
High Binding
Polystyrene
Non-sterile
Yes
100/Cs.
127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.)
High Binding
8 x 12
Clear, Blue
96
Curved Edges, Flat Bottom (C-well)
Colorimetric immunoassay
Yes
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions