missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates

Catalog No. 07000779
Change view
Click to view available options
Color:
Blue, Clear Bottom
Clear
Clear, Blue
Clear, Green
Clear, Red
Clear, Yellow
Green, Clear Bottom
Red, Clear Bottom
Binding Property:
High Binding
Medium Binding
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Binding Property
07000779 Clear Medium Binding
07000775 Red, Clear Bottom Medium Binding
07000777 Green, Clear Bottom Medium Binding
07000776 Blue, Clear Bottom Medium Binding
07000780 Clear High Binding
07000781 Clear, Red High Binding
07000782 Clear, Blue High Binding
07000783 Clear, Green High Binding
07000784 Clear, Yellow High Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 07000779 Supplier Greiner Bio-One Supplier No. 705070

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Individual wells can be pipetted without leaving residue
  • Flat bottom enables precise optical measurements
  • Suitable for transmission measurements
  • Specifica

Service Plan

No

Warranty

No

Specifications

Breakapart Single break
Binding Property Medium Binding
Format Strip Plate
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon
Well Shape Curved Edges, Flat Bottom (C-well)
Dnase Free Yes
For Use With (Application) Colorimetric immunoassay
Quantity 100/Cs.
RNase Free Yes
Dimensions (L x W x H) 127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.)
Show More Show Less