Learn More
DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial Caps with PTFE/Silicone Septa
Available in multiple colors and package sizes.
$88.84 - $2,430.86
Specifications
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
View More Specs
Products 23
Catalog Number Mfr. No. Color Septum Pre-slit No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Color Septum Pre-slit No. per Pack Price Quantity & Availability  
15111066
View Documents Promotion Details
DWK Life Sciences
090030B
Blue PTFE, Silicone No 100
Pack of 100 Each for $88.84
Only null left
15111067
View Documents Promotion Details
DWK Life Sciences
090030BB
Blue PTFE, Silicone No 2000
Pack of 2000 Each for $1,775.88
Only null left
15111069
View Documents Promotion Details
DWK Life Sciences
090030G
Green PTFE, Silicone No 100
Pack of 100 Each for $88.84
Only null left
15111070
View Documents Promotion Details
DWK Life Sciences
090030GB
Green PTFE, Silicone No 2000
Pack of 2000 Each for $1,775.88
Only null left
15111072
View Documents Promotion Details
DWK Life Sciences
090030K
Black PTFE, Silicone No 100
Pack of 100 Each for $88.84
Only null left
15111073
View Documents Promotion Details
DWK Life Sciences
090030KB
Black PTFE, Silicone No 2000
Pack of 2000 Each for $1,811.40
Only null left
15111075
View Documents Promotion Details
DWK Life Sciences
090030N
Natural PTFE, Silicone No 100
Pack of 100 Each for $88.84
Only null left
15111076
View Documents Promotion Details
DWK Life Sciences
090030NB
Natural PTFE, Silicone No 2000
Pack of 2000 Each for $1,775.88
Only null left
15111078
View Documents Promotion Details
DWK Life Sciences
090030R
Red PTFE, Silicone No 100
Pack of 100 Each for $88.84
Only null left
15111079
View Documents Promotion Details
DWK Life Sciences
090030RB
Red PTFE, Silicone No 2000
Pack of 2000 Each for $1,775.88
Only null left
15111081
View Documents Promotion Details
DWK Life Sciences
090030Y
Yellow PTFE, Silicone No 100
Pack of 100 Each for $88.84
Only null left
15111082
View Documents Promotion Details
DWK Life Sciences
090030YB
Yellow PTFE, Silicone No 2000
Pack of 2000 Each for $1,775.88
Only null left
15111086
View Documents Promotion Details
DWK Life Sciences
090035B
Blue PTFE, Silicone Yes 100
Pack of 100 Each for $135.96
Only null left
15111087
View Documents Promotion Details
DWK Life Sciences
090035BB
Blue PTFE, Silicone Yes 2000
Pack of 2000 Each for $2,407.20
Only null left
15111089
View Documents Promotion Details
DWK Life Sciences
090035G
Green PTFE, Silicone Yes 100
Pack of 100 Each for $150.47
Only null left
15111090
View Documents Promotion Details
DWK Life Sciences
090035GB
Green PTFE, Silicone Yes 2000
Pack of 2000 Each for $2,175.10
Only null left
15111091
View Documents Promotion Details
DWK Life Sciences
090035K
Black PTFE, Silicone Yes 100
Pack of 100 Each for $150.47
Only null left
15111092
View Documents Promotion Details
DWK Life Sciences
090035KB
Black PTFE, Silicone Yes 2000
Pack of 2000 Each for $2,430.86
Only null left
15111093
View Documents Promotion Details
DWK Life Sciences
090035N
Natural PTFE, Silicone Yes 100
Pack of 100 Each for $150.47
Only null left
15111096
View Documents Promotion Details
DWK Life Sciences
090035R
Red PTFE, Silicone Yes 100
Pack of 100 Each for $135.96
Only null left
15111097
View Documents Promotion Details
DWK Life Sciences
090035RB
Red PTFE, Silicone Yes 2000
Pack of 2000 Each for $2,175.10
Only null left
15111098
View Documents Promotion Details
DWK Life Sciences
090035Y
Yellow PTFE, Silicone Yes 100
Pack of 100 Each for $150.47
Only null left
15111100
View Documents Promotion Details
DWK Life Sciences
090036B
Blue Bonded PTFE, Silicone Yes 100
Pack of 100 Each for $140.84
Only null left
Description
Description
Specifications
Specifications
MicroLiter
9 mm
9 mm Autosampler Vials - MicroLiter