Learn More
DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial Caps with PTFE/Silicone Septa
Available in multiple colors and package sizes.
Supplier: DWK Life Sciences 090030BB
Chemical Identifiers
| MicroLiter | |
| 9 mm | |
| 9 mm Autosampler Vials - MicroLiter |
Specifications
| Blue | |
| No | |
| 2000 | |
| 9 mm | |
| Screw Cap |
| Polypropylene | |
| PTFE, Silicone | |
| MicroLiter | |
| 9 mm Autosampler Vials - MicroLiter |