missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial Caps with PTFE/Silicone Septa

Catalog No. 15111100
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
Septum:
Bonded PTFE, Silicone
PTFE, Silicone
Pre-slit:
No
Yes
No. per Pack:
100
2000
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Pre-slit No. per Pack
15111100 Blue Bonded PTFE, Silicone Yes 100
15111066 Blue PTFE, Silicone No 100
15111067 Blue PTFE, Silicone No 2000
15111069 Green PTFE, Silicone No 100
15111070 Green PTFE, Silicone No 2000
15111072 Black PTFE, Silicone No 100
15111073 Black PTFE, Silicone No 2000
15111075 Natural PTFE, Silicone No 100
15111076 Natural PTFE, Silicone No 2000
15111078 Red PTFE, Silicone No 100
15111079 Red PTFE, Silicone No 2000
15111081 Yellow PTFE, Silicone No 100
15111082 Yellow PTFE, Silicone No 2000
15111086 Blue PTFE, Silicone Yes 100
15111087 Blue PTFE, Silicone Yes 2000
15111089 Green PTFE, Silicone Yes 100
15111090 Green PTFE, Silicone Yes 2000
15111091 Black PTFE, Silicone Yes 100
15111092 Black PTFE, Silicone Yes 2000
15111093 Natural PTFE, Silicone Yes 100
15111096 Red PTFE, Silicone Yes 100
15111097 Red PTFE, Silicone Yes 2000
15111098 Yellow PTFE, Silicone Yes 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 15111100 Supplier DWK Life Sciences Supplier No. 090036B

Available in multiple colors and package sizes.

Specifications

Color Blue
Material Polypropylene
Pre-slit Yes
Septum Bonded PTFE, Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap