missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers

Catalog No. 15111722
Change view
Click to view available options
Color:
Black
Gray
White, Tan
Material:
Butyl
PTFE, Silicone
Product Type:
3-leg Stopper
Septa
Stopper
No. per Pack:
100
1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Product Type No. per Pack
15111722 White, Tan PTFE, Silicone Septa 1000
15111657 Black Butyl Septa 100
15111670 Gray Butyl Stopper 100
15111671 Gray Butyl Stopper 1000
15111672 Gray Butyl 3-leg Stopper 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15111722 Supplier DWK Life Sciences Supplier No. 200051B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers are designed for use with 20 mm crimp seal headspace vials. Butyl stoppers suit applications where PTFE is not required; PTFE/butyl and molded PTFE/butyl septa deliver low background peaks and a tight seal.

  • Designed for use with WHEATON MicroLiter 20 mm crimp seal headspace vials
  • Butyl-only and butyl stopper options available for applications where PTFE is not required
  • PTFE/butyl septa provide an economical choice with low background peaks
  • Molded PTFE/butyl septa offer the tightest seal of all PTFE items and low background peaks
  • PTFE/silicone septa are excellent for volatile compound analysis
  • Aluminum foil/silicone septa are suitable for volatile applications
  • Multiple material combinations available to match instrument and application requirements
  • Available in standard packs of 100 and bulk packs of 1000

Specifications

Color White, Tan
Diameter (Metric) Outer 20 mm
Material PTFE, Silicone
Pre-slit No
Sterility Non-sterile
No. per Pack 1000
Quantity 1000/Cs.
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Seals - MicroLiter
Autoclavable Autoclavable
Product Type Septa
Show More Show Less