missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20 mm Aluminum Crimp Top Headspace Vial Seals, No Liner

Catalog No. 15111629
Change view
Click to view available options
Color:
Blue
Red
Silver
No. per Pack:
100
1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
15111629 Blue 100
15111641 Red 100
15111627 Silver 100
15111628 Silver 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15111629 Supplier DWK Life Sciences Supplier No. 200000B

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

Specifications

Color Blue
Material Aluminum
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Septa - MicroLiter
Product Type Crimp Cap