missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microlite™ White Microtiter™ Plates
GREENER_CHOICE

Catalog No. 14245181
Change view
Click to view available options
Product Type:
Microplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14245181 Microplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14245181 Supplier Thermo Scientific™ Supplier No. 741712

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Microlite Microtiter Plates.

Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Thermo Scientific™ Microlite White Microtiter Plates.

Available in plates or 12-well breakable strips.

  • Polystyrene

     

  • Choice of two surfaces, each performance assayed using an IgG binding assay

SURFACES

  • Microlite 1+ - Hydrophobic surface for binding of biomolecules with hydrophilic domains

  • Microlite 2+ - Slightly hydrophilic surface for increased binding of biomolecules with hydrophilic/hydrophobic character such as many proteins

Note:

Other configurations are available in Microlite 1 and 2 formulations.

Specifications

Binding Property Slightly Hydrophilic
Format Standard
Description Plate; Microlite 2
Detection Method Chemiluminescence
Array 8 x 12
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlite 1+
Plate Blocking None
Quantity Each
Target Molecule Antibodies (Glycoproteins) and Proteins
Show More Show Less