missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ MICROCHEM™ by AlphaTec™ Breathable Oversleeves

Catalog No. 19500751
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19500751 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19500751 Supplier Ansell™ Supplier No. WH20B9260000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ansell™ MICROCHEM™ by AlphaTec™ Breathable Oversleeves feature microporous laminate technology

  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
  • Free of natural rubber latex
  • Applications:
    • Clean room cleaning and preparing
    • Production line support and maintenance
    • Maintenance of plant and machines
    • Veterinary services

Specifications

Product Type Oversleeves
Color White
Material Non-woven Microporous Polyethylene Laminate
Size Universal