missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates

Catalog No. A36924
Encompass
Change view
Click to view available options
Quantity:
20 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36924 20 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A36924 Supplier Applied Biosystems™ Supplier No. A36924

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs.

MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs. EnduraPlate™ plastic consumables are validated with Applied Biosystems™ instruments for optimal fit and performance. They are constructed to ANSI/SBS standards and are well-suited for automated workflows. MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates are semi-skirted and fit standard (0.2 mL) 96-well thermal blocks.

  • Optimized for performance with Applied Biosystems instruments
  • Certified DNA-, RNase-, and PCR inhibitor-free
  • Constructed to ANSI/SBS standards for compatibility with high-throughput laboratories and robotic handling
  • Easy-to-read black well-identification text to minimize pipetting errors

EnduraPlate™ reaction plates use polypropylene for a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from the thermal block to the sample, while a polycarbonate frame provides mechanical stability, flatness, and durability that is compatible with automation/robotics. Easy-to-read, black well-identification text helps to minimize pipetting mistakes and improves sample preparation.

MicroAmp EnduraPlate™ Optical 96-Well Clear Reaction Plates are provided without barcodes, enabling customization with your own barcodes

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Store at room temperature.
Packaging 20 Plates
Product Line MicroAmp
Color Optical, Clear
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polycarbonate, Polypropylene
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 20 plates
Shipping Condition Room Temperature
Show More Show Less