missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Micro pH Electrodes

pH electrodes designed for small sample volumes or for measuring in small containers.

$761.47 - $1,535.10

Specifications

Type Combination Electrode
Response Time <20 sec.
View More Specs

Products 6
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
01911477
Accessories View Documents Promotion Details
METTLER TOLEDO™
51343160
60 mm
Each for $821.90
Only null left
 
01911476
View Documents Promotion Details
METTLER TOLEDO™
51343161
230 mm
Each for $761.47
Only null left
 
01911475
View Documents Promotion Details
METTLER TOLEDO™
51343165
100 mm
Each for $1,045.50
Only null left
 
01917147
View Documents Promotion Details
METTLER TOLEDO™
51344163
130 mm
Each for $1,535.10
Only null left
 
01911442
View Documents Promotion Details
METTLER TOLEDO™
51343164
180 mm
Each for $1,404.81
Only null left
 
01914306
View Documents Promotion Details
METTLER TOLEDO™
30244732
40 mm
Each for $1,025.10
Only null left
 
Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. (Connection cable is not included; sold separately.)
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Vials
  • Microplates
  • Test tubes
  • Serum and Gastric juice
  • Micro-biological samples
  • Paint
  • Gel
  • Soap
  • Shampoo
  • Cosmetics

Electrode Specifications

  • InLab Micro: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Micro Pro-ISM: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Ultra-Micro-ISM: ceramic junction; refillable with FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Semi-Micro: ceramic junction; non-refillable - replace when electrolyte depleted
  • InLab Semi-Micro-L: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Flex-Micro: porous PTFE junction; non-refillable - replace when electrolyte depleted

Accessories & Care

  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

Combination Electrode
<20 sec.
Videos
SDS
Documents

Documents