missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Micro pH Electrodes

pH electrodes designed for small sample volumes or for measuring in small containers.

Supplier:  METTLER TOLEDO™ 51343161

 View more versions of this product

Catalog No. 01911476


Only null left

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. (Connection cable is not included; sold separately.)
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Vials
  • Microplates
  • Test tubes
  • Serum and Gastric juice
  • Micro-biological samples
  • Paint
  • Gel
  • Soap
  • Shampoo
  • Cosmetics

Electrode Specifications

  • InLab Micro: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Micro Pro-ISM: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Ultra-Micro-ISM: ceramic junction; refillable with FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Semi-Micro: ceramic junction; non-refillable - replace when electrolyte depleted
  • InLab Semi-Micro-L: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Flex-Micro: porous PTFE junction; non-refillable - replace when electrolyte depleted

Accessories & Care

  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Chemical Identifiers

Combination Electrode
<20 sec.

Specifications

S7
Glass
0°C to 100°C
3mol/L KCl
Requires Cable, S7 to BNC (30281915)
U Glass
<20 sec.
32°F to 212°F
9.05 in.
0.23 in.
Ceramic
0 to 14
Combination Electrode
Measure pH in the small or narrow neck vessels
ARGENTHAL with Ag+ trap
InLab™ Semi-Micro-L
3mol/L KCl
0°C to 100°C
230 mm
6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions