missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ Micro pH Electrodes
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. (Connection cable is not included; sold separately.)
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Vials
- Microplates
- Test tubes
- Serum and Gastric juice
- Micro-biological samples
- Paint
- Gel
- Soap
- Shampoo
- Cosmetics
Electrode Specifications
- InLab Micro: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Micro Pro-ISM: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Ultra-Micro-ISM: ceramic junction; refillable with FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
- InLab Semi-Micro: ceramic junction; non-refillable - replace when electrolyte depleted
- InLab Semi-Micro-L: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Flex-Micro: porous PTFE junction; non-refillable - replace when electrolyte depleted
Accessories & Care
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Specifications
| Connector Types | Multi-Pin |
| Junction | Ceramic |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | Combination Electrode |
| Filling Solution Type | 3mol/L KCl |
| For Use With (Application) | Measure pH in the smallest vessels used in the laboratory |
| For Use With (Equipment) | Requires Cable, Multi-Pin to BNC/RCA (30281896) |
| Includes | ISM Temperature Probe |
| Show More |